Merck
Confirmation of co-denitrification in grazed grassland.
Scientific reports (2015-12-01)
Diana R Selbie, Gary J Lanigan, Ronald J Laughlin, Hong J Di, James L Moir, Keith C Cameron, Tim J Clough, Catherine J Watson, James Grant, Cathal Somers, Karl G Richards
RESUMEN
Pasture-based livestock systems are often associated with losses of reactive forms of nitrogen (N) to the environment. Research has focused on losses to air and water due to the health, economic and environmental impacts of reactive N. Di-nitrogen (N2) emissions are still poorly characterized, both in terms of the processes involved and their magnitude, due to financial and methodological constraints. Relatively few studies have focused on quantifying N2 losses in vivo and fewer still have examined the relative contribution of the different N2 emission processes, particularly in grazed pastures. We used a combination of a high (15)N isotopic enrichment of applied N with a high precision of determination of (15)N isotopic enrichment by isotope-ratio mass spectrometry to measure N2 emissions in the field. We report that 55.8 g N m(-2) (95%, CI 38 to 77 g m(-2)) was emitted as N2 by the process of co-denitrification in pastoral soils over 123 days following urine deposition (100 g N m(-2)), compared to only 1.1 g N m(-2) (0.4 to 2.8 g m(-2)) from denitrification. This study provides strong evidence for co-denitrification as a major N2 production pathway, which has significant implications for understanding the N budgets of pastoral ecosystems.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cloruro de potasio, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Cloruro de potasio, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Cloruro de potasio, Molecular Biology, ≥99.0%
Sigma-Aldrich
Cloruro de potasio, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Cloruro de potasio, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Cloruro de potasio, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Cloruro de potasio solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Cloruro de potasio, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Dicyandiamide, 99%
Sigma-Aldrich
Cloruro de potasio, BioXtra, ≥99.0%
Sigma-Aldrich
Cloruro de potasio, 99.999% trace metals basis
Sigma-Aldrich
Cloruro de potasio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de potasio, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Cloruro de potasio solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Cloruro de potasio, ≥99.99% trace metals basis
Sigma-Aldrich
Cloruro de potasio, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cloruro de potasio, AnhydroBeads, −10 mesh, 99.99% trace metals basis