Merck
A unique homodimeric NAD⁺-linked isocitrate dehydrogenase from the smallest autotrophic eukaryote Ostreococcus tauri.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-03-01)
Wang-Gang Tang, Ping Song, Zheng-Yu Cao, Peng Wang, Guo-Ping Zhu
RESUMEN
In eukaryotes, NAD(+)-dependent isocitrate dehydrogenase (IDH) is strictly mitochondrial and is a key enzyme in the Krebs cycle. To date, all known NAD(+)-specific IDHs (NAD-IDHs) in the mitochondria are believed to be heteromeric in solution. Here, a unique homodimeric NAD-IDH from Ostreococcus tauri (OtIDH), the smallest autotrophic picoeukaryote, was unveiled. Active OtIDH has a molecular weight of ∼93 kDa with each subunit of 46.7 kDa. In the presence of Mn(2+) and Mg(2+), OtIDH displayed 42-fold and 51-fold preference for NAD(+) over NADP(+), respectively. Interestingly, OtIDH exhibited a sigmoidal kinetic behavior in response to isocitrate unlike other homodimeric homologs, and a remarkably high affinity for isocitrate (S0.5 < 10 μM) unlike other hetero-oligomeric homologs. Furthermore, its coenzyme specificity can be completely converted from NAD(+) (ancient trait) to NADP(+) (adaptive trait) by rational mutagenesis based on the evolutionary trace. Mutants D344R and D344R/M345H displayed a 15-fold and 72-fold preference for NADP(+) over NAD(+), respectively, indicating that D344 and M345 are the determinants of NAD(+) specificity. These findings also suggest that OtIDH may be an ancestral form of type II IDHs (all reported members are NADP(+)-linked enzymes) and may have evolved into NADP(+)-dependent IDH for adaptation to the increased demand of NADPH under carbon starvation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Glicerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Glicerol, ≥99.5%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Glicerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Glicerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Glicerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glicerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Sigma-Aldrich
Glicerol, FCC, FG
Sigma-Aldrich
Glicerol, meets USP testing specifications
Sigma-Aldrich
Sulfato de sodio, ≥99.99% trace metals basis
Sigma-Aldrich
Sulfato de sodio, BioXtra, ≥99.0%
Sigma-Aldrich
Cloruro de sodio, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sulfato de sodio, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Cloruro de sodio, tablet
Sigma-Aldrich
Cloruro de sodio, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Cloruro de sodio, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sulfato de sodio, ≥99.0%, suitable for plant cell culture