Merck
A solid-contact ion selective electrode for copper(II) using a succinimide derivative as ionophore.
Sensors (Basel, Switzerland) (2013-04-04)
Mihaela Dana Tutulea-Anastasiu, Deivy Wilson, Manel del Valle, Cristina Mihaela Schreiner, Igor Cretescu
RESUMEN
All-solid-state sensors with polyvinyl chloride (PVC)-based membranes using off-the-shelf N-hydroxysuccinimide (NHS) and succinimide (Succ) ionophores were prepared using DOP (dioctyl phthalate) and NPOE (ortho-nitrophenyloctyl ether) as plasticizers. Good responses were obtained when NHS was used. The potentiometric response of the proposed electrode is independent of pH over the range 2-6. The electrode shows a fast response time of 0.25 s. The electrode exhibits a Super-Nernstian response, with 37.5 mV/decade, with a potentiometric detection limit of 4.4 µM. The proposed sensor revealed good selectivity towards a group of transition metal ions.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Potassium tetrakis(4-chlorophenyl)borate, Selectophore, ≥98.0%