Merck
Histone acetylation regulates orphan nuclear receptor NR4A1 expression in hypercholesterolaemia.
Clinical science (London, England : 1979) (2015-09-24)
Xina Xie, Xuhong Song, Song Yuan, Haitao Cai, Yequn Chen, Xiaolan Chang, Bin Liang, Dongyang Huang
RESUMEN
Hypercholesterolaemia and inflammation are correlated with atherogenesis. Orphan nuclear receptor NR4A1, as a key regulator of inflammation, is closely associated with lipid levels in vivo. However, the mechanism by which lipids regulate NR4A1 expression remains unknown. We aimed to elucidate the underlying mechanism of NR4A1 expression in monocytes during hypercholesterolaemia, and reveal the potential role of NR4A1 in hypercholesterolaemia-induced circulating inflammation. Circulating leucocytes were collected from blood samples of 139 patients with hypercholesterolaemia and 139 sex- and age-matched healthy subjects. We found that there was a low-grade inflammatory state and higher expression of NR4A1 in patients. Both total cholesterol and low-density lipoprotein cholesterol levels in plasma were positively correlated with NR4A1 mRNA level. ChIP revealed that acetylation of histone H3 was enriched in the NR4A1 promoter region in patients. Human mononuclear cell lines THP-1 and U937 were treated with cholesterol. Supporting our clinical observations, cholesterol enhanced p300 acetyltransferase and decreased HDAC7 (histone deacetylase 7) recruitment to the NR4A1 promoter region, resulting in histone H3 hyperacetylation and further contributing to NR4A1 up-regulation in monocytes. Moreover, cytosporone B, an NR4A1 agonist, completely reversed cholesterol-induced IL-6 (interleukin 6) and MCP-1 (monocyte chemoattractant protein 1) expression to below basal levels, and knockdown of NR4A1 expression by siRNA not only mimicked, but also exaggerated the effects of cholesterol on inflammatory biomarker up-regulation. Thus we conclude that histone acetylation contributes to the regulation of NR4A1 expression in hypercholesterolaemia, and that NR4A1 expression reduces hypercholesterolaemia-induced inflammation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cytosporone B, ≥98% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
MISSION® esiRNA, targeting human NR4A1
En este momento no podemos mostrarle ni los precios ni la disponibilidad