Merck
Dependence of brown adipose tissue function on CD36-mediated coenzyme Q uptake.
Cell reports (2015-01-27)
Courtney M Anderson, Melissa Kazantzis, Jinshan Wang, Subramaniam Venkatraman, Renata L S Goncalves, Casey L Quinlan, Ryan Ng, Martin Jastroch, Daniel I Benjamin, Biao Nie, Candice Herber, An-Angela Ngoc Van, Michael J Park, Dawee Yun, Karen Chan, Angela Yu, Peter Vuong, Maria Febbraio, Daniel K Nomura, Joseph L Napoli, Martin D Brand, Andreas Stahl
RESUMEN
Brown adipose tissue (BAT) possesses the inherent ability to dissipate metabolic energy as heat through uncoupled mitochondrial respiration. An essential component of the mitochondrial electron transport chain is coenzyme Q (CoQ). While cells synthesize CoQ mostly endogenously, exogenous supplementation with CoQ has been successful as a therapy for patients with CoQ deficiency. However, which tissues depend on exogenous CoQ uptake as well as the mechanism by which CoQ is taken up by cells and the role of this process in BAT function are not well understood. Here, we report that the scavenger receptor CD36 drives the uptake of CoQ by BAT and is required for normal BAT function. BAT from mice lacking CD36 displays CoQ deficiency, impaired CoQ uptake, hypertrophy, altered lipid metabolism, mitochondrial dysfunction, and defective nonshivering thermogenesis. Together, these data reveal an important new role for the systemic transport of CoQ to BAT and its function in thermogenesis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Reactivo glicerol libre, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Coenzyme Q10, ≥98% (HPLC)