Merck
Rhizopus stolonifer mediated biosynthesis of biocompatible cadmium chalcogenide quantum dots.
Enzyme and microbial technology (2016-11-22)
P Mareeswari, J Brijitta, S Harikrishna Etti, C Meganathan, Gobi Saravanan Kaliaraj
RESUMEN
We report an efficient method to biosynthesize biocompatible cadmium telluride and cadmium sulphide quantum dots from the fungus Rhizopus stolonifer. The suspension of the quantum dots exhibited purple and greenish-blue luminescence respectively upon UV light illumination. Photoluminescence spectroscopy, X-ray diffraction, and transmission electron microscopy confirms the formation of the quantum dots. From the photoluminescence spectrum the emission maxima is found to be 424 and 476nm respectively. The X-ray diffraction of the quantum dots matches with results reported in literature. The 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay for cell viability evaluation carried out on 3-days transfer, inoculum 3×10
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tellurium tetrachloride, 99%