Merck
Ibuprofen Induces Mitochondrial-Mediated Apoptosis Through Proteasomal Dysfunction.
Molecular neurobiology (2015-12-17)
Arun Upadhyay, Ayeman Amanullah, Deepak Chhangani, Vibhuti Joshi, Ribhav Mishra, Amit Mishra
RESUMEN
In routine course of life, nonsteroidal anti-inflammatory drugs (NSAIDs) are widely prescribed antipyretic, analgesic, and anti-inflammatory drugs. It is a well-proposed notion that treatment of NSAIDs may induce anti-proliferative effects in numerous cancer cells. Ibuprofen from isobutylphenylpropanoic acid is NSAID and used to relieve fever, pain, and inflammation. It is also used for juvenile idiopathic arthritis, rheumatoid arthritis, patent ductus arteriosus, and for pericarditis. Despite few emerging studies have expanded the fundamental concept that the treatment of NSAIDs influences apoptosis in cancer cells, however the NSAID-mediated precise mechanisms that determine apoptosis induction without producing adverse consequences in variety of cancer cells are largely unknown. In our present study, we have observed that ibuprofen reduces proteasome activity, enhances the aggregation of ubiquitylated abnormal proteins, and also elevates the accumulation of crucial proteasome substrates. Ibuprofen treatment causes mitochondrial abnormalities and releases cytochrome c into cytosol. Perhaps, the more detailed study is needed in the future to elucidate the molecular mechanisms of NSAIDs that can induce apoptosis without adverse effects and produce effective anti-tumor effects and consequently help in neurodegeneration and ageing.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)