Merck
  • Dexamethasone preventing contractile and cytoskeletal protein changes in the rabbit basilar artery after subarachnoid hemorrhage.
Dexamethasone preventing contractile and cytoskeletal protein changes in the rabbit basilar artery after subarachnoid hemorrhage.
Journal of neurosurgery (2005-05-06)
Philippe Gomis, Yves Roger Tran-Dinh, Christine Sercombe, Richard Sercombe
RESUMEN
The aim of this project was to study the perturbations of four smooth-muscle proteins and an extracellular protein, type I collagen, after subarachnoid hemorrhage (SAH) and to examine the possible preventive effects of dexamethasone. Using a one-hemorrhage rabbit model, the authors first examined the effects of SAH on the expression of alpha-actin, h-caldesmon, vimentin, smoothelin-B, and type I collagen; second, they studied whether post-SAH systemic administration of dexamethasone (three daily injections) corrected the induced alterations. Measurements were obtained at Day 7 post-SAH. The proteins were studied by performing immunohistochemical staining and using a laser-scanning confocal microscope. Compared with control (sham-injured) arteries, the density of the media of arteries subjected to SAH was reduced for alpha-actin (-11%, p = 0.01) and h-caldesmon (-15%, p = 0.06) but increased for vimentin (+15%, p = 0.04) and smoothelin-B (+53%, p = 0.04). Among animals in which SAH was induced, arteries in those treated with dexamethasone demonstrated higher values of density for alpha-actin (+13%, p = 0.05) and h-caldesmon (+20%, p = 0.01), lower values for vimentin (-55%, p = 0.05), and nonsignificantly different values for smoothelin-B. The density of type I collagen in the adventitia decreased significantly after SAH (-45%, p = 0.01), but dexamethasone treatment had no effect on this decrease. The SAH-induced alterations in the density of three of four smooth-muscle proteins were prevented by dexamethasone treatment; two of these proteins--alpha-actin and h-caldesmon--are directly related to contraction. This drug may potentially be useful to prevent certain morphological and functional changes in cerebral arteries after SAH.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Smoothelin Antibody, clone R4A, clone R4A, Chemicon®, from mouse