Merck
Analytical study of penicillamine in pharmaceuticals by capillary zone electrophoresis.
Journal of chromatography. A (2000-01-15)
R Gotti, R Pomponio, V Andrisano, V Cavrini
RESUMEN
The ability of capillary zone electrophoresis in the development of analytical methods devoted to the quality control of the thiol drug penicillamine is shown. Using 50 mM phosphate running buffer (pH 2.5), good quantitations of underivatized penicillamine and its disulfide were achieved; detection at 200 nm allowed checking the presence of the disulfide impurity in pharmaceuticals. The use of 1,1-[ethenylidenebis(sulfonyl)]bis-benzene as a thiol specific reagent resulted in an increased sensitivity for the quantitation of D-penicillamine (limit of detection at 200 nm wavelength was 1.5 microM). Introducing beta-cyclodextrin as chiral selector in the running buffer, enantioseparation of D-L-penicillamine was obtained; for this purpose (+)-camphor-10-sulfonic acid, a chiral ion-pairing reagent, was found to be an essential additive in obtaining a baseline separation. The resulting enantioseparative system was validated in order to evaluate the presence of the toxic L-penicillamine enantiomer in pharmaceutical samples.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
D-Penicillamine, 98-101%