Merck
Zinc alpha2 glycoprotein alleviates palmitic acid-induced intracellular lipid accumulation in hepatocytes.
Molecular and cellular endocrinology (2016-06-07)
Xinhua Xiao, Han Li, Xiaoyan Qi, Yadi Wang, Canxin Xu, Gexin Liu, Gebo Wen, Jianghua Liu
RESUMEN
Zinc alpha2 glycoprotein (ZAG) plays an important role in stimulating fat mobilization and lipolysis in adipose tissue, but its role in hepatic lipid metabolism remains unclear. Palmitic acid (PA) was used to stimulate HepG2 cells with ZAG overexpression or ZAG knock down (shRNA). Overexpression of ZAG significantly inhibited lipogenesis, promoted lipolysis and fatty acid β-oxidation, and attenuated PA-induced intracellular fat accumulation. Moreover, ZAG overexpression dramatically stimulated adiponectin expression in HepG2 cells. In contrast, knockdown of ZAG notably inhibited fatty acid β-oxidation, increased lipogenesis and lipid accumulation. Collectively, these data suggest that ZAG has the potential to alleviate hepatosteatosis, making it a promising therapeutic target for fatty liver.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human AZGP1