Merck
  • Ribonucleic acid interference knockdown of IL-6 enhances the efficacy of cisplatin in laryngeal cancer stem cells by down-regulating the IL-6/STAT3/HIF1 pathway.
Ribonucleic acid interference knockdown of IL-6 enhances the efficacy of cisplatin in laryngeal cancer stem cells by down-regulating the IL-6/STAT3/HIF1 pathway.
Cancer cell international (2017-09-08)
Qiang Fu, Pengruofeng Liu, Xiumei Sun, Shanshan Huang, Fengchan Han, Lili Zhang, Yannan Xu, Tingyan Liu
RESUMEN
Cisplatin has been used in the treatment of many cancers, including laryngeal cancer; however, its efficacy can be reduced due to the development of drug resistance. This study aimed to investigate whether interleukin-6 (IL-6) knockdown may enhance the efficacy of cisplatin in laryngeal cancer stem cells (CSC) and the potential involvement of the signal transducer and activator of transcription 3 (STAT3) and hypoxia-inducible factor 1 (HIF1) in this effect. The ALDH+ and CD44+ CSC in Hep2 human laryngeal squamous cancer cells were identified by the fluorescence-activated cell sorting technique. IL-6, STAT3 and HIF1 mRNA and protein expressions were examined with quantitative real-time polymerase chain reaction and Western blot, respectively. Cell proliferation was measured by MTT assay. Tumorigenicity was measured by a colony formation assay and invasion was determined by a cell invasion assay. Apoptotic cells were counted by flow cytometry. Immunohistochemistry was performed to detect immunoreactive IL-6, STAT3 and HIF1 cells in xenografts. The mRNA and protein levels of IL-6, STAT3 and HIF1 were significantly increased in Hep2-CSC as compared with those from Hep2 cells. Application of siRNA-IL-6 to knockdown IL-6 resulted in significantly decreased IL-6, STAT3 and HIF1 mRNA and protein levels. IL-6 knockdown reduced cell proliferation, tumorigenicity and invasion and increased apoptosis within CSC. Enhanced degrees of suppression in these parameters were observed when IL-6 knockdown was combined with cisplatin in these CSC. Results from the xenograft study showed that the combination of IL-6 knockdown and cisplatin further inhibited the growth of xenografts as compared with that obtained in the cisplatin-injected group alone. Immunoreactive IL-6, STAT3 and HIF1 cell numbers were markedly reduced in IL-6 knockdown tumor tissues. IL-6, STAT3 and HIF1 immunoreactive cell counts were further reduced in tissue where IL-6 knockdown was combined with cisplatin treatment as compared with tissue receiving cisplatin alone. IL-6 knockdown can increase chemo-drug efficacy of cisplatin, inhibit tumor growth and reduce the potential for tumor recurrence and metastasis in laryngeal cancer. The IL-6/STAT3/HIF1 pathway may represent an important target for investigating therapeutic strategies for the treatment of laryngeal cancer.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human IL6