Merck
Research on Controllable Degradation of Novel Sulfonylurea Herbicides in Acidic and Alkaline Soils.
Journal of agricultural and food chemistry (2017-08-17)
Shaa Zhou, Xue-Wen Hua, Wei Wei, Yu-Cheng Gu, Xiao-Qing Liu, Jing-Huo Chen, Ming-Gui Chen, Yong-Tao Xie, Sha Zhou, Xiang-De Meng, Yan Zhang, Yong-Hong Li, Bao-Lei Wang, Hai-Bin Song, Zheng-Ming Li
RESUMEN
The degradation issue of sulfonylurea (SU) has become one of the biggest challenges that hamper the development and application of this class of herbicides, especially in the alkaline soils of northern China. On the basis of the previous discovery that some substituents on the fifth position of the benzene ring in Chlorsulfuron could hasten its degradation rate, apparently in acidic soil, this work on Metsulfuron-methyl showed more convincing results. Two novel compounds (I-1 and I-2) were designed and synthesized, and they still retained potent herbicidal activity in tests against both dicotyledons and monocotyledons. The half-lives of degradation (DT
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1,3-Propylene sulfite, 99%