Merck
  • Antitumor and antimetastatic activities of rhamnogalacturonan-II-type polysaccharide isolated from mature leaves of green tea via activation of macrophages and natural killer cells.
Antitumor and antimetastatic activities of rhamnogalacturonan-II-type polysaccharide isolated from mature leaves of green tea via activation of macrophages and natural killer cells.
International journal of biological macromolecules (2017-02-23)
Hye-Ryung Park, Dahyun Hwang, Hyung-Joo Suh, Kwang-Won Yu, Tae Young Kim, Kwang-Soon Shin
RESUMEN
To investigate the antitumor and antimetastatic polysaccharide from the mature leaves of green tea, GTE-II was purified using size exclusion chromatography. GTE-II consisted of 15 different sugars including rarely observed sugars such as 2-O-methyl-fucose, 2-O-methyl-xylose, apiose, aceric acid, 3-deoxy-d-manno-2-octulosonic acid, and 3-deoxy-d-lyxo-2-heptulosaric acid, which were characteristics of pectic polysaccharide rhamnogalacturonan-II. Treatment of peritoneal macrophages with GTE-II not only increased interleukin (IL)-6 and IL-12 production, but also had significantly increased tumoricidal activity against Yac-1 tumor cells than those obtained from untreated mice. In an assay of natural killer (NK) cell activity, intravenous administration of GTE-II significantly stimulated NK cytotoxicity against Yac-1 tumor cells. Furthermore, the depletion of NK cells by injection of rabbit anti-asialo GM1 serum eliminated the inhibitory effect of GTE-II on B16BL6 melanoma cells. These data suggest that GTE-II inhibits tumor metastasis, and its antitumor effect is associated with activation of macrophages and NK cells.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
3-Phenylphenol, 85%