Merck
(Electro)chemical Oxidation of 6,13-Bis[tri(isopropyl)silylethynyl]pentacene to its Radical Cation and Dication.
Chemphyschem : a European journal of chemical physics and physical chemistry (2017-06-08)
Simon Schundelmeier, Bernd Speiser, Holger F Bettinger, Ralf Einholz
RESUMEN
6,13-Bis[tri(isopropyl)silylethynyl]pentacene is a prototypical molecule for organic semiconductor and photovoltaic materials, which makes its electrochemical (redox) properties highly interesting. However, previous cyclic voltammetric studies have provided only limited information. Kinetic and persistence information and identification of the oxidation product(s) and their further reaction or oxidation have not been reported. Thus, an extended electrochemical and spectroscopic investigation of this compound was conducted in CH
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ferrocene, 98%