Merck
Amplified gene location in chromosomal DNA affected recombinant protein production and stability of amplified genes.
Biotechnology progress (2000-10-12)
T Yoshikawa, F Nakanishi, Y Ogura, D Oi, T Omasa, Y Katakura, M Kishimoto, K Suga
RESUMEN
Previously, we established an easy and quick construction method for obtaining a stable and highly productive gene-amplified recombinant Chinese hamster ovary (CHO) cell line. With a gradual increase in methotrexate (MTX) concentration, gene-amplified cell pools had high and stable specific growth and production rates. Moreover, the phenotype of gene-amplified cells seemed to be affected by the location of the amplified gene in chromosomal DNA. We suspected that various kinds of gene-amplified cells might appear during the long-term selection to construct gene-amplified cell pools. To clarify the behavior of gene-amplified cell pools during a stepwise increase of MTX concentration, we isolated gene-amplified clones derived from gene-amplified cell pools. We compared the characteristics of isolated clones, such as the productivity of recombinant protein, stability of amplified genes, and the location of amplified genes. As a result, telomere-type clones, in which the amplified gene was located near the telomeric region, were found to be more stable and productive than other types of clones. Telomere-type clones had over 100 copies of amplified genes in the chromosomal DNA. In contrast, a large number of other types of clones had less than 10 copies of amplified genes. During long-term cultivation in the absence of MTX, in other types of clones, amplified genes rapidly decreased in the chromosomal DNA.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dihydrofolic acid, ≥90%
En este momento no podemos mostrarle ni los precios ni la disponibilidad