Merck
Firing of Replication Origins Frees Dbf4-Cdc7 to Target Eco1 for Destruction.
Current biology : CB (2017-09-19)
Agustin I Seoane, David O Morgan
RESUMEN
Robust progression through the cell-division cycle depends on the precisely ordered phosphorylation of hundreds of different proteins by cyclin-dependent kinases (CDKs) and other kinases. The order of CDK substrate phosphorylation depends on rising CDK activity, coupled with variations in substrate affinities for different CDK-cyclin complexes and the opposing phosphatases [1-4]. Here, we address the ordering of substrate phosphorylation by a second major cell-cycle kinase, Cdc7-Dbf4 or Dbf4-dependent kinase (DDK). The primary function of DDK is to initiate DNA replication by phosphorylating the Mcm2-7 replicative helicase [5-7]. DDK also phosphorylates the cohesin acetyltransferase Eco1 [8]. Sequential phosphorylations of Eco1 by CDK, DDK, and Mck1 create a phosphodegron that is recognized by the ubiquitin ligase SCF
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Concanamycin A, ≥70% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad