Merck
  • Phase I dose-escalation study of the c-Met tyrosine kinase inhibitor SAR125844 in Asian patients with advanced solid tumors, including patients with
Phase I dose-escalation study of the c-Met tyrosine kinase inhibitor SAR125844 in Asian patients with advanced solid tumors, including patients with
Oncotarget (2017-11-08)
Kohei Shitara, Tae Min Kim, Tomoya Yokota, Masahiro Goto, Taroh Satoh, Jin-Hee Ahn, Hyo Song Kim, Sylvie Assadourian, Corinne Gomez, Marzia Harnois, Satoshi Hamauchi, Toshihiro Kudo, Toshihido Doi, Yung-Jue Bang
RESUMEN
SAR125844 is a potent and selective inhibitor of the c-Met kinase receptor. This was an open-label, phase I, multicenter, dose-escalation, and dose-expansion trial of SAR125844 in Asian patients with solid tumors, a subgroup of whom had gastric cancer and
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Mecoprop, PESTANAL®, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad