Merck
Autophosphorylation-based Calcium (Ca
The Journal of biological chemistry (2017-02-06)
Kyle W Bender, R Kevin Blackburn, Jacqueline Monaghan, Paul Derbyshire, Frank L H Menke, Cyril Zipfel, Michael B Goshe, Raymond E Zielinski, Steven C Huber
RESUMEN
Plant calcium (Ca
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-polihistidina monoclonal antibody produced in mouse, clone HIS-1, ascites fluid