Merck
Epigenetic silencing of
Oncotarget (2018-02-13)
Hironori Aoki, Eiichiro Yamamoto, Akira Takasawa, Takeshi Niinuma, Hiro-O Yamano, Taku Harada, Hiro-O Matsushita, Kenjiro Yoshikawa, Ryo Takagi, Eiji Harada, Yoshihito Tanaka, Yuko Yoshida, Tomoyuki Aoyama, Makoto Eizuka, Akira Yorozu, Hiroshi Kitajima, Masahiro Kai, Norimasa Sawada, Tamotsu Sugai, Hiroshi Nakase, Hiromu Suzuki
RESUMEN
Colorectal sessile serrated adenoma/polyps (SSA/Ps) are well-known precursors of colorectal cancer (CRC) characterized by
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-β-actina monoclonal antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Sigma-Aldrich
MISSION® esiRNA, targeting human SMOC1