Merck
Characterization of cyclic adenosine diphosphate-ribose-induced Ca2+ release in T lymphocyte cell lines.
Journal of immunology (Baltimore, Md. : 1950) (1995-10-01)
A H Guse, C P da Silva, F Emmrich, G A Ashamu, B V Potter, G W Mayr
RESUMEN
Ca2+ release from intracellular stores is one of the major events transducing extracellular signals into living cells. Recently, a metabolite of nicotinamide adenine dinucleotide+ (NAD+), termed "cyclic adenosine diphosphate-ribose" (cADPr), has been described to release Ca2+ from caffeine-sensitive internal stores of cells. Jurkat T cells possess intracellular Ca2+ stores sensitive to caffeine, so a potential involvement of cADPr in Ca2+ signaling was investigated. cADPr released Ca2+ in a dose-dependent manner from intracellular stores of permeabilized Jurkat T cells. Half maximal release was obtained at 2.25 microM cADPr. Prior addition of D-myo-inositol 1,4,5-trisphosphate (Ins(1,4,5)P3) or thapsigargin did not influence cADPr-induced Ca2+ release, indicating the presence of different Ca2+ pools sensitive to Ins(1,4,5)P3 and cADPr. The specificity of the response was confirmed using the inhibitors ruthenium red, 8-NH2-cADPr, and 8-Br-cADPr. All three compounds blocked cADPr-induced, but not Ins(1,4,5)P3-induced, Ca2+ release in a dose-dependent manner. Cyclic GMP (cGMP)-induced Ca2+ release was also partly antagonized by ruthenium red, indicating involvement of a cGMP-dependent step in the formation of cADPr. The presence of endogenous cADPr was analyzed directly by HPLC. Sequential separation on strong anion exchange HPLC and reverse-phase, ion-pair HPLC resulted in a single symmetric peak co-eluting with standard cADPr. The identity of this endogenous material was further confirmed by its ability to release Ca2+ in saponin-permeabilized Jurkat T cells.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
8-Bromo-cyclic adenosine diphosphate ribose, 85% (HPLC), lyophilized powder