Detection and Differentiation of Staphylococcus Aureus on Baird Parker Agar
Jvo Siegrist
Product Manager Microbiology,
AnalytiX Volume 10 Article 4
Baird Parker Agar is used for the isolation and differentiation of coagulase-positive staphylococci in food and pharmaceuticals according to the Baird Parker method.
As a nitrogen source for the organism, casein peptone and meat extract are added to the medium. Yeast extract provides nitrogen as well as other important nutrients like vitamin B12 complex. The medium contains lithium and tellurite, which inhibit most of the contaminating microflora, while glycine and pyruvate enhance Staphylococci growth. Staphylococci can reduce tellurite to telluride, which results in grey to black coloration of the colonies. With the addition of egg yolk, the medium becomes yellow and slightly opaque. A clear halo develops around colonies from coagulase-positive Staphylococcus aureus, and upon further incubation may produce an opaque zone due to an egg yolk–lecithinase reaction (lipolytic activity). Grey-black colonies and a halo on this medium are presumed to be indicative of coagulase-positive staphylococci, as a high correlation between coagulase test and lipophilic activity was found. Staphylococcus aureus and some strains of Staphylococcus saprophyticus (Shaw et al.[5]) may show both of these characteristics, but they are easily distinguished from each other by the different times at which the halo develops.
Baird Parker Agar Base
Baird Parker Agar with Rabbit Plasma Fibrinogen can be used for the detection of coagulase activity. Coagulase-positive organisms appear as grey to black colonies because of the tellurite reduction and as an opaque halo, due to the conversion by coagulase from fibrinogen to fibrin. Only plates with less than 100 characteristic colonies should be counted. The use of this medium eliminates the need for an additional coagulase test.
Media:
Supplements:
- Egg yolk tellurite emulsion (Cat. No. 75208)
- RPF Supplement (Rabbit Plasma Fibrinogen Supplement; Cat. No 05939)
- Egg-Yolk Tellurite Emulsion Sterile, 20% 1.03785.0001
Microorganisms for testing performance:
| Material No. | Productivity species (all concentrations) | WDCM No. | CFU |
|---|---|---|---|
| CRM06571L-10EA | Staphylococcus aureus | 00035 | 30-120 |
| CRM06571M-10EA | Staphylococcus aureus | 00035 | 500-50,000 |
| VT000357-10EA | Staphylococcus aureus | 00035 | 50,000-150,000 |
| VT000322-10EA | Staphylococcus aureus subsp. aureus | 00032 | 15-80 |
| VT000323-10EA | Staphylococcus aureus subsp. aureus | 00032 | 80-130 |
| VT000324-10EA | Staphylococcus aureus subsp. aureus | 00032 | 130-300 |
| VT000326-10EA | Staphylococcus aureus subsp. aureus | 00032 | 1,000-10,000 |
| VT000327-10EA | Staphylococcus aureus subsp. aureus | 00032 | 50,000-150,000 |
| VT000342-10EA | Staphylococcus aureus subsp. aureus | 00034 | 15-80 |
| VT000343-10EA | Staphylococcus aureus subsp. aureus | 00034 | 80-130 |
| VT000346-10EA | Staphylococcus aureus subsp. aureus | 00034 | 1,000-10,000 |
| VT000347-10EA | Staphylococcus aureus subsp. aureus | 00034 | 50,000-150,000 |
| Material No. | Selectivity species | WDCM No. | CFU |
|---|---|---|---|
| VT000127-10EA | Escherichia coli | 00012 | 50,000-150,000 |
| VT000137-10EA | Escherichia coli | 00013 | 50,000-150,000 |
| Material No. | Specificity species | WDCM No. | CFU |
|---|---|---|---|
| VT001596-10EA | Staphylococcus saprophyticus | 00159 | 1,000-10,000 |