Inchi Code
1S/C3H4BrN3/c4-2-1-6-7-3(2)5/h1H,(H3,5,6,7)
Inchi Key
OELYMZVJDKSMOJ-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Keep in dark place, sealed in dry, room temperature
Country of Origin
CN
Physical Form
Pale-yellow to brown or green or gray powder or crystals
Still not finding the right product?
Explore all of our products under 4-Bromo-1H-pyrazol-3-amine
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
