Skip to Content
MilliporeSigma

BDPH9BCC7D65

Bide Pharmatech

6-Fluoro-2-aminophenol

Synonym(s):

2-amino-6-fluorophenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6O1N1F1
CAS Number:
Molecular Weight:
127.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6FNO/c7-4-2-1-3-5(8)6(4)9/h1-3,9H,8H2

Inchi Key

MDIAVSZFIQWYST-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Yellow to Brown or Gray Solid



Still not finding the right product?

Explore all of our products under 6-Fluoro-2-aminophenol


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library