Skip to Content
MilliporeSigma

BDPH9BCD1280

Bide Pharmatech

1-Butylquinoline-2,4(1H,3H)-dione

Synonym(s):

1-butyl-4-hydroxy-2(1H)-quinolinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H15O2N1
CAS Number:
Molecular Weight:
217.27
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H15NO2/c1-2-3-8-14-11-7-5-4-6-10(11)12(15)9-13(14)16/h4-7H,2-3,8-9H2,1H3

Inchi Key

VBGFLYWUTPWXPK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 1-Butylquinoline-2,4(1H,3H)-dione



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library