Skip to Content
MilliporeSigma

BDPH9C0220EF

Bide Pharmatech

Kp7-6

Synonym(s):

(S)-2-((((4R,7S,10S,13S,16S,19R)-10-((1H-imidazol-5-yl)methyl)-19-(((S)-2-amino-1-hydroxy-3-(4-hydroxyphenyl)propylidene)amino)-7-benzyl-13-(2-carboxyethyl)-16-(carboxymethyl)-6,9,12,15,18-pentahydroxy-1,2-dithia-5,8,11,14,17-pentaazacycloicosa-5,8,11,14,17-pentaen-4-yl)(hydroxy)methylene)amino)-3-(4-hydroxyphenyl)propanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C48H56N10O15S2/c49-31(16-26-6-10-29(59)11-7-26)41(65)57-37-22-74-75-23-38(47(71)56-36(48(72)73)18-27-8-12-30(60)13-9-27)58-43(67)33(17-25-4-2-1-3-5-25)53-44(68)34(19-28-21-50-24-51-28)54-42(66)32(14-15-39(61)62)52-45(69)35(20-40(63)64)55-46(37)70/h1-13,21,24,31-38,59-60H,14-20,22-23,49H2,(H,50,51)(H,52,69)(H,53,68)(H,54,66)(H,55,70)(H,56,71)(H,57,65)(H,58,67)(H,61,62)(H,63,64)(H,72,73)/t31-,32-,33-,34-,35-,36-,37-,38-/m0/s1

Inchi Key

FXIRLIREUIBHCF-QSVFAHTRSA-N

Shipping Temperature

Cold-chain transportation

Purity

99%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under Kp7-6



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library