Direkt zum Inhalt
Merck

A013-M

NovaSeptum® Single-sampling Unit Bag-Rack

For use with 5-port holder, TC 50.5 for NAC-USI, NAC-USM

Synonym(e):

single-sample bag rack, sterile sample bag rack

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern

Über diesen Artikel

eCl@ss:
36101790
UNSPSC Code:
41104105
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


material

stainless steel rack

Quality Segment

sterility

non-sterile

product line

NovaSeptum®

packaging

pkg of 1 unit

L × W × H

250 mm (9.8 in.) × 280 mm (11 in.) × 260-460 mm (10.2-18.1 in.)

weight

0.5 kg (1.1 lb)

application(s)

sterile sampling

compatibility

for use with 5-port holder, TC 50.5 for NAC-USI, NAC-USM

General description

The NovaSeptum® Bag Rack for Single Sampling Units is a stainless steel mechanical support rack designed to hold and organize NovaSeptum® sampling units during aseptic fluid sampling operations. The rack is engineered for compatibility with the 5-port TC 50.5 (Tri-Clamp 50.5 mm) holders compatible with NAC-USI and NAC-USM (NovAseptic Connector) connectors. The rack is fully adjustable, reusable, and available for all standard holders.

Application

  • The NovaSeptum® Bag Rack acts as a structured mechanical support frame that holds and organizes NovaSeptum® sampling units during aseptic fluid sampling operations.
  • It is particularly suited for use in biopharmaceutical manufacturing and cleanroom environments where controlled, contamination-free fluid sampling is required.

Features and Benefits

  • Fully adjustable to accommodate different process setups.
  • The maximum load capacity of the rack goes up to 5 kgs.
  • The structural components are manufactured from stainless steel in compliance with ASTM® 316, while the screws and locking nuts are made from A4-grade stainless steel.
  • A certificate of quality is included in each package for traceability and compliance.
  • Compatible with bag, bottle and conical tube sampling units

Packaging

A complete bag rack unit, including all components, is packaged unassembled in a cardboard box and assigned a part number.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

Die passende Version wird nicht angezeigt?

Wenn Sie eine bestimmte Version benötigen, können Sie anhand der Lot- oder Chargennummer nach einem spezifischen Zertifikat suchen.

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen