Direkt zum Inhalt
Merck

AB5470

Anti-Potassium Channel TASK-2 Antibody

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern

Über diesen Artikel

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


conjugate

unconjugated

Quality Segment

antibody form

affinity purified immunoglobulin

clone

polyclonal

species reactivity

rat, rabbit

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

General description

FUNCTION: SwissProt: O95279 # pH-dependent, voltage insensitive, outwardly rectifying potassium channel. Outward rectification is lost at high external K(+) concentrations.
SIZE: 499 amino acids; 55130 Da
SUBUNIT: Homodimer; disulfide-linked.
SUBCELLULAR LOCATION: Membrane; Multi-pass membrane protein (Potential).
TISSUE SPECIFICITY: Abundant expression in kidney, also detected in liver, placenta and small intestine. In the kidney, expression is restricted to the distal tubules and collecting ducts. Not expressed in proximal tubules or glomeruli.
SIMILARITY: SwissProt: O95279 ## Belongs to the two pore domain potassium channel (TC 1.A.1.8) family.
MISCELLANEOUS: Inhibited by quinine, quinidine and external acidification.
A purified peptide (TASK-2) corresponding to amino acid residues 483-499 of human TASK-2 (Accession O95279).

Application

Western blot: 1.5 μg/mL (1:200) on rat kidney membranes using ECL.

Immunohistochemistry on rat kidney tissue sections.

Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

The epitope is specific for TASK-2 and is not present in any other known proteins.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Preparation Note

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

CONTROL ANTIGEN: Included free of charge with the antibody is 20 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Lagerklasse

11 - Combustible Solids



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

Die passende Version wird nicht angezeigt?

Wenn Sie eine bestimmte Version benötigen, können Sie anhand der Lot- oder Chargennummer nach einem spezifischen Zertifikat suchen.

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen