Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Species reactivity:
human
Technique(s):
western blot: suitable
Application:
WB
Citations:
-
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenQuality Segment
biological source
rabbit
antibody form
purified antibody
antibody product type
primary antibodies
mol wt
calculated mol wt 15.40 kDa, observed mol wt ~16 kDa
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
mouse, rat
packaging
antibody small pack of 100
technique(s)
western blot: suitable
isotype
IgG
epitope sequence
N-terminal
Protein ID accession no.
UniProt accession no.
storage temp.
2-8°C
Gene Information
human ... H3C1(8350)
General description
Histone H3.1 (UniProt: P68431; also known as Histone H3/a, 3/b,3/c, 3/d, 3/e, 3/f, 3/h, 3/i, 3/j, 3/k, 3/l) is encoded by the H3C1 gene (Gene ID: 8350) in human. Histones are highly conserved basic nuclear proteins that are responsible for the nucleosome structure of chromatin in eukaryotes. They play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. The amino-terminal tails of histone proteins are subject to posttranslational modifications, including acetylation and methylation, which recruit downstream regulatory factors, influence chromatin structure, and are critical determinants of transcription. Histone H3Q5amphetaminylation is a novel post-translational modification, where the glutamine-5 residue of histone H3 becomes covalently modified by amphetamine (a central nervous system stimulant which is commonly used to treat ADHD and narcolepsy) via transglutaminase 2 (TG2)-mediated incorporation. This modification is structurally similar to histone dopaminylation, which has been implicated in transcriptional regulation and neuronal function. Given the psychoactive properties of amphetamine and it′s long-term use by patients, H3Q5 amphetaminylation could in theory influence gene expression patterns in a drug-specific manner, potentially affecting neuronal plasticity, addiction-related pathways, and other neuronal processes. Characterization of this modification could provide insight into how amphetamines modify long-term epigenetics in the brain and body, with implications for neuropsychiatric disorders, substance use research, and more.
Immunogen
KLH-conjugated linear peptide corresponding to 10 amino acids from the N-terminal region of human Histone H3.1 modified with amphetamine at glutamine 5 position.
Application
Quality Control Testing
Evaluated by Western Blotting in Recombinant TG2, Recombinant mononucleosomes reacted with amphetamine.
Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected Histone H3.1 modified with amphetamine at Glutamine 5, in Recombinant TG2 + Recombinant mononucleosomes reacted with 5mM amphetamine.
Evaluated by Western Blotting in Recombinant TG2, Recombinant mononucleosomes reacted with amphetamine.
Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected Histone H3.1 modified with amphetamine at Glutamine 5, in Recombinant TG2 + Recombinant mononucleosomes reacted with 5mM amphetamine.
Biochem/physiol Actions
This rabbit polyclonal antibody specifically detects Histone H3.1 modified with amphetamine at glutamine 5 position.
Physical form
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
0.25 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommended storage: +2°C to +8°C.
Analysis Note
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen
Lagerklasse
12 - Non Combustible Liquids
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.