Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 mg | Warenkorb auf Verfügbarkeit prüfen | € 308,00 |
Über diesen Artikel
biological source
goat
Quality Segment
conjugate
TRITC conjugate, rhodamine conjugate
antibody form
affinity purified immunoglobulin
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable, immunofluorescence: suitable
shipped in
wet ice
target post-translational modification
unmodified
General description
Immunogen
Application
ELISA
Optimal working dilutions must be determined by the end user.
Secondary & Control Antibodies
Secondary Antibodies Adsorbed for Dual Labeling
Fragment Specific Secondary Antibodies
Biochem/physiol Actions
Physical form
Preparation Note
Legal Information
Disclaimer
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Chemicon®, from goat | description affinity isolated antibody, lyophilized powder | description affinity isolated antibody, lyophilized powder | description affinity isolated antibody, lyophilized powder |
| antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| biological source goat | biological source goat | biological source rabbit | biological source goat |
| technique(s) ELISA: suitable, immunofluorescence: suitable | technique(s) direct immunofluorescence: suitable, indirect immunofluorescence: suitable, indirect ELISA: suitable | technique(s) direct immunofluorescence: suitable, indirect immunofluorescence: suitable, indirect ELISA: suitable | technique(s) indirect ELISA: suitable, direct immunofluorescence: suitable, indirect immunofluorescence: suitable |
| species reactivity human | species reactivity human | species reactivity goat | species reactivity rabbit |
| conjugate TRITC conjugate, rhodamine conjugate | conjugate rhodamine conjugate | conjugate rhodamine conjugate | conjugate rhodamine conjugate |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Still not finding the right product?
Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen
Was ist los?
WGK 2
Lagerklasse
12 - Non Combustible Liquids
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



