Anmelden zur Ansicht der Organisations- und Vertragspreise.
GröĂe auswĂ€hlen
Ansicht Àndern
Ăber diesen Artikel
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
26, monoclonal
Species reactivity:
mouse, human
Application:
IHC, WB
Citations:
-
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist fĂŒr Sie da.
UnterstĂŒtzung erhaltenGeneral description
FUNCTION: SwissProt: P02686 # The classic group of MBP isoforms (isoform 4-isoform 14) are with PLP the most abundant protein components of the myelin membrane in the CNS. They have a role in both its formation and stabilization. The smaller isoforms might have an important role in remyelination of denuded axons in multiple sclerosis. The non- classic group of MBP isoforms (isoform 1-isoform 3/Golli-MBPs) may preferentially have a role in the early developing brain long before myelination, maybe as components of transcriptional complexes, and may also be involved in signaling pathways in T- cells and neural cells. Differential splicing events combined to optional post-translational modifications give a wide spectrum of isomers, each of them having maybe a specialized function. Induces T-cell proliferation.
SIZE: 304 amino acids; 33117 Da
SUBUNIT: Homodimer; isoform 3 exists as a homodimer.
SUBCELLULAR LOCATION: Myelin membrane; Peripheral membrane protein; Cytoplasmic side. Note=Cytoplasmic side of myelin.
TISSUE SPECIFICITY: MBP isoforms are found in both the central and the peripheral nervous system, whereas Golli-MBP isoforms are expressed in fetal thymus, spleen and spinal cord, as well as in cell lines derived from the immune system.DEVELOPMENTAL STAGE: Expression turns on abruptly in fetus of 14 to 16 weeks. Even smaller isoforms seem to be produced during embryogenesis, some of these persisting in the adult. Expression of isoform MBP2 is more evident at 16 weeks and its relative proportion declined thereafter.
PTM: Several charge isomers of MBP; C1 (the most cationic, least modified, and most abundant form), C2, C3, C4, C5, C6, C7, C8-A and C8-B (the least cationic form); are produced as a result of optional PTM, such as phosphorylation, deamidation of glutamine or asparagine, arginine citrullination and methylation. C8-A and C8-B contain each two mass isoforms termed C8-A(H), C8-A(L), C8-B(H) and C8-B(L), (H) standing for higher and (L) for lower molecular weight. C3, C4 and C5 are phosphorylated. The ratio of methylated arginine residues decreases in aging, making the protein more cationic. & The N-terminal alanine is acetylated (isoform 3, isoform 4, isoform 5 and isoform 6). & Arg-241 was found to be 6% monomethylated and 60% symmetrically dimethylated.
DISEASE: SwissProt: P02686 # The reduction in the surface charge of citrullinated and/or methylated MBP could result in a weakened attachment to the myelin membrane. This mechanism could be operative in demyelinating diseases such as chronical multiple sclerosis (MS), and fulminating MS (Marburg disease).
SIMILARITY: SwissProt: P02686 ## Belongs to the myelin basic protein family.
SIZE: 304 amino acids; 33117 Da
SUBUNIT: Homodimer; isoform 3 exists as a homodimer.
SUBCELLULAR LOCATION: Myelin membrane; Peripheral membrane protein; Cytoplasmic side. Note=Cytoplasmic side of myelin.
TISSUE SPECIFICITY: MBP isoforms are found in both the central and the peripheral nervous system, whereas Golli-MBP isoforms are expressed in fetal thymus, spleen and spinal cord, as well as in cell lines derived from the immune system.DEVELOPMENTAL STAGE: Expression turns on abruptly in fetus of 14 to 16 weeks. Even smaller isoforms seem to be produced during embryogenesis, some of these persisting in the adult. Expression of isoform MBP2 is more evident at 16 weeks and its relative proportion declined thereafter.
PTM: Several charge isomers of MBP; C1 (the most cationic, least modified, and most abundant form), C2, C3, C4, C5, C6, C7, C8-A and C8-B (the least cationic form); are produced as a result of optional PTM, such as phosphorylation, deamidation of glutamine or asparagine, arginine citrullination and methylation. C8-A and C8-B contain each two mass isoforms termed C8-A(H), C8-A(L), C8-B(H) and C8-B(L), (H) standing for higher and (L) for lower molecular weight. C3, C4 and C5 are phosphorylated. The ratio of methylated arginine residues decreases in aging, making the protein more cationic. & The N-terminal alanine is acetylated (isoform 3, isoform 4, isoform 5 and isoform 6). & Arg-241 was found to be 6% monomethylated and 60% symmetrically dimethylated.
DISEASE: SwissProt: P02686 # The reduction in the surface charge of citrullinated and/or methylated MBP could result in a weakened attachment to the myelin membrane. This mechanism could be operative in demyelinating diseases such as chronical multiple sclerosis (MS), and fulminating MS (Marburg disease).
SIMILARITY: SwissProt: P02686 ## Belongs to the myelin basic protein family.
Human myelin basic protein.
Application
Epitope: a.a. 67-74
Biochem/physiol Actions
Reacts with an epitope of myelin basic protein between aa 67-74.
Amino acid sequence YGSLPQ. Reacts with MBP from most species.
Amino acid sequence YGSLPQ. Reacts with MBP from most species.
Physical form
Tissue culture supernatant. Liquid containing 0.09% sodium azide.
Preparation Note
Maintain at -20° in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze-thaw cycles.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-Myelin Basic Protein Antibody, a.a. 67-74, clone 26
Lagerklasse
12 - Non Combustible Liquids
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kĂŒrzlich erworben haben.