Direkt zum Inhalt
Merck

MABC1810

Anti-Globo H ceramide/GHCer Antibody, clone VK9

Synonym(e):

Globo H ceramide

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern

Über diesen Artikel

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Clone:
VK9, monoclonal
Species reactivity:
human
Application:
DB, ELISA, EM, FACS, ICC, IF, IHC, IP, inhibition assay
Citations:
-
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

VK9, monoclonal

mol wt

calculated mol wt N/A kDa, observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable, dot blot: suitable, electron microscopy: suitable, flow cytometry: suitable, immunocytochemistry: suitable, immunofluorescence: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, inhibition assay: suitable

isotype

IgG3κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Immunogen

Globo H-KLH conjugate

Application

Quality Control Testing

Evaluated by Immunohistochemistry in human liver cancer tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Globo H-ceramide (GHCer) in human liver cancer tissue sections.

Tested Applications

Inhibition Assay: A representative lot of this antibody inhibited blood vessel formation in Matrigel plug and reduced hemoglobulin accumulation. (Wang, S-H., et al. (2022). J Biomed Sci. 29:105).

Electron Microscopy: A representative lot detected Globo H ceramide/GHCer in Electron Microscopy application (Wang, S-H., et al. (2022). J Biomed Sci. 29:105).

Dot Blot: A representative lot detected Globo H ceramide/GHCer in Dot Blot application (Wang, S-H., et al. (2022). J Biomed Sci. 29:105).

Immunoprecipitation Analysis: A representative lot immunoprecipitated Globo H ceramide/GHCer in Immunoprecipitation application (Kudryashov, V., et al. (1998). Glycoconj J. 15(3):243-9; Wang, S-H., et al. (2022). J Biomed Sci. 29:105).

Immunofluorescence Analysis: A representative lot detected Globo H ceramide/GHCer in Immunofluorescence application (Lo, T-C., et al. (2023). Dev Cell. 58(22):2447-2459).

Flow Cytometry Analysis: A representative lot detected Globo H ceramide/GHCer in Flow Cytometry application (Lo, T-C., et al. (2023). Dev Cell. 58(22):2447-2459).

Immunocytochemistry Analysis: A representative lot detected Globo H ceramide/GHCer in Immunocytochemistry application (Kudryashov, V., et al. (1998). Glycoconj J. 15(3):243-9).

ELISA: A representative lot detected Globo H ceramide/GHCer in an ELISA application (Kudryashov, V., et al. (1998). Glycoconj J. 15(3):243-9).

Immunohistochemistry Analysis: A representative lot detected Globo H ceramide/GHCer in Immunohistochemistry application (Wang, S-H., et al. (2022). J Biomed Sci. 29:105).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone VK9 is a mouse monoclonal antibody that specifically detects Globo H-ceramide (GHCer).

Physical form

Purified mouse monoclonal antibody IgG3 in PBS without preservatives.

Preparation Note

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Glycosphingolipids (GSLs) are a heterogeneous class of membrane glycolipids consisting of a ceramide backbone covalently linked to a glycan moiety and are involved in many cellular processes. Globo H (Fuc 1 2Galβ1 3GalNAcβ13Gal 14Galβ1 4Glc) is a carbohydrate structure, which is synthesized by specific glycosyltransferases, and shows enhanced expression in many human carcinomas. Globo H-ceramide (GHCer) is a glycosphingolipid and consists of a ceramide backbone with a carbohydrate chain that includes the Globo H, and serves as a critical tumor-associated carbohydrate antigen. It is predominantly expressed in various epithelial cancers, including breast, pancreatic, and colorectal cancers. GHCer is involved in cell signaling pathways that regulate tumor progression, immune evasion, and angiogenesis. Its expression is notably upregulated in malignant cells compared to normal tissues, making it a potential biomarker for cancer diagnosis and prognosis. Subcellularly, Globo H-ceramide localizes in lipid rafts and membrane microdomains, facilitating interactions with signaling molecules and receptors. Its involvement in disease is highlighted by its role in promoting cancer cell proliferation and metastasis, as well as its potential as a target for immunotherapeutic strategies aimed at enhancing anti-tumor immunity. This mAb VK-9 is suitable for multiple applications including western blotting, immunocytochemistry, immunoprecipitation, flow cytometry ELISA, and inhibition studies. It does not cross react with many related structures, such as galactosylgloboside, globoside, H type 1, H type 2 blood group structures or fucosyl-gangliotetraosyl ceramide, but reacts weakly with globo A ceramide. It recognizes globo H-ceramide and human tumor cells expressing globo H. (Ref.: Kudryashov, V., et al. (1998). Glycoconj J. 15(3):243-9; Wang, S-H., et al. (2022). J Biomed Sci. 29:105; Lo, T-C., et al. (2023). Dev Cell. 58(22):2447-2459.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen


Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable

Lagerklasse

12 - Non Combustible Liquids



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

Die passende Version wird nicht angezeigt?

Wenn Sie eine bestimmte Version benötigen, können Sie anhand der Lot- oder Chargennummer nach einem spezifischen Zertifikat suchen.

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen