Direkt zum Inhalt
Merck

Fortfahren mit

MABF435

Anti-CD45.2 Antibody (mouse), APC, clone 104

clone 104, 0.2 mg/mL, from mouse

Synonym(e):

Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, Lymphocyte antigen 5, Ly-5, T200, CD45

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
Allophycocyanin conjugate
Clone:
104, monoclonal
Application:
FACS, ICC, IP
Citations:
-
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

mouse

Quality Segment

conjugate

Allophycocyanin conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

104, monoclonal

species reactivity

mouse

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ptprc(19264)

General description

The 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to specific for CD45.2 and is not cross-reactive with CD45.1-bearing cells.

Application

This mouse monoclonal antibody Anti-CD45.2 (mouse), APC, clone 104 validated for use in IP, Flow Cytometry & ICC for the detection of CD45.2.

Analysis Note

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.5 μg of this antibody detected CD45.2 in one million C57Bl/6 splenocytes.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
MABF598MABF438MABF439
description

clone 104, 0.2 mg/mL, from mouse

description

clone 104, 0.2 mg/mL, from mouse

description

clone 104, 0.2 mg/mL, from mouse

description

clone 104, 0.2 mg/mL, from mouse

conjugate

Allophycocyanin conjugate

conjugate

PE

conjugate

PE

conjugate

PE-Cy7

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

Gene Information

mouse ... Ptprc(19264)

Gene Information

mouse ... Ptprc(19264)

Gene Information

mouse ... Ptprc(19264)

Gene Information

mouse ... Ptprc(19264)

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

purified antibody

clone

104, monoclonal

clone

104, monoclonal

clone

104, monoclonal

clone

104, monoclonal


Still not finding the right product?

Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen


Lagerklasse

12 - Non Combustible Liquids

WGK

nwg



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen




Questions

Reviews

No rating value

Active Filters