Merck
MABS150
Anti-HGK Antibody, clone 2H1.1
Größe auswählen
Ansicht ändern
Über diesen Artikel
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
2H1.1, monoclonal
Application:
WB
Citations:
-
Anmelden – hier finden Sie Ihre Preise
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
conjugate
unconjugated
Quality Segment
antibody form
purified antibody
clone
2H1.1, monoclonal
purified by
using protein G
species reactivity
mouse, rat
concentration
(Please refer to lot specific datasheet.)
technique(s)
western blot: suitable
isotype
IgG2aκ
UniProt accession no.
target post-translational modification
unmodified
General description
Recombinant protein corresponding to human HGK.
~150 kDa observed
Physical form
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in C6 cell lysate.
Western Blotting Analysis: 1.0 µg/mL of this antibody detected HGK in 10 µg of C6 cell lysate.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)