Über diesen Artikel
NACRES:
NB.75
UNSPSC Code:
41122409
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenpackaging
pkg of 1 ea
manufacturer/tradename
Milliflex Oasis®
technique(s)
Rapid Sterility Testing: suitable
application(s)
bioburden testing
pharmaceutical
sterility testing
compatibility
for use with Milliflex Oasis® VHP vacuum pump
General description
Milliflex Oasis® filtration system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. Milliflex Oasis® VHP pump is a new generation vacuum pump for sterility and bioburden analysis in pharmaceutical environments. The Milliflex Oasis® VHP electrical cable is used for connecting two pumps together and benefit from a high throughput workflow using one single power supply.
Application
Milliflex Oasis® VHP vacuum pump finds its application in bioburden testing and sterility testing for Pharmaceutical Products Analysis.
Legal Information
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?