Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
UNSPSC Code:
41106514
NACRES:
NA.81
eCl@ss:
32040201
Biological source:
human
Relevant disease(s):
cancer
Technique(s):
cell culture | mammalian: suitable
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenbiological source
human
Quality Segment
description
cell culture mammalian: suitable
packaging
vial of ≥1X10⁶ vial (viable cells per vial)
manufacturer/tradename
Millipore
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
liquid nitrogen
storage temp.
−196°C
General description
Soft tissue sarcomas (STS) are a rare type of tumor made up of mixed cell populations with mesenchymal features. They are classified into many histological subtypes such as undifferentiated pleomorphic sarcoma (UPS). UPS is a subtype previously known as malignant fibrous histiocytoma. UPS is associated with one of the worst prognoses of STSs and is frequently diagnosed as a high-grade tumor. Metastatic forms of this cancer have a very low 5-year survival rate of about 16%. There is also still little understanding of the UPS subtype which involves various cellular backgrounds and mutations.
Immunotherapy continues to be a strong candidate for cancer treatment in recent years. It has already shown strong results for certain cancer types that did not see effective treatment with previously traditional treatment methods. While the immunotherapy options for STS can be limited due to tumor complexity, the UPS subtype appears to show encouraging results from immunotherapy, specifically with PD-1/PD-L1 interactions. However, there is still limited availability of patient-derived UPS cell lines which has constrained the amount of preclinical research conducted so far.
The SCC649 JBT-19 undifferentiated pleomorphic sarcoma line is a strong option for continuing UPS research. The JBT-19 cell line appears to be the first introduced UPS cell line to simultaneously express PD-L1 and collagen, two important ligands of known immune checkpoint molecules, PD-1 and LAIR-1. This makes the cell line a strong candidate for immunotherapeutic research. The cell line can grow in both 2D and 3D formats in vitro as well as being able to produce solid tumors in mice when inoculated in vivo.
The JBT-19 cell line was established from a solid UPS tumor of a 65-year-old male. A combination of tumor fragmentation-dissociation techniques was used to establish the cells in an adherent monolayer.
Source
Cells were established from a solid UPS (undifferentiated pleomorphic sarcoma) primary tumor of a 65-year-old male diagnosed with the disease. Tumor was surgically removed, and the cell line was established in monolayer using a combination of tumor fragmentation dissociation techniques.
References
1. Taborska P, Lukac P, Stakheev D, Rajsiglova L, Kalkusova K, Strnadova K, Lacina L, Dvorankova B, Novotny J, Kolar M, et al. 2023. Novel PD-L1- and collagen-expressing patient-derived cell line of undifferentiated pleomorphic sarcoma (JBT19) as a model for cancer immunotherapy. Scientific Reports. 13(1).
Immunotherapy continues to be a strong candidate for cancer treatment in recent years. It has already shown strong results for certain cancer types that did not see effective treatment with previously traditional treatment methods. While the immunotherapy options for STS can be limited due to tumor complexity, the UPS subtype appears to show encouraging results from immunotherapy, specifically with PD-1/PD-L1 interactions. However, there is still limited availability of patient-derived UPS cell lines which has constrained the amount of preclinical research conducted so far.
The SCC649 JBT-19 undifferentiated pleomorphic sarcoma line is a strong option for continuing UPS research. The JBT-19 cell line appears to be the first introduced UPS cell line to simultaneously express PD-L1 and collagen, two important ligands of known immune checkpoint molecules, PD-1 and LAIR-1. This makes the cell line a strong candidate for immunotherapeutic research. The cell line can grow in both 2D and 3D formats in vitro as well as being able to produce solid tumors in mice when inoculated in vivo.
The JBT-19 cell line was established from a solid UPS tumor of a 65-year-old male. A combination of tumor fragmentation-dissociation techniques was used to establish the cells in an adherent monolayer.
Source
Cells were established from a solid UPS (undifferentiated pleomorphic sarcoma) primary tumor of a 65-year-old male diagnosed with the disease. Tumor was surgically removed, and the cell line was established in monolayer using a combination of tumor fragmentation dissociation techniques.
References
1. Taborska P, Lukac P, Stakheev D, Rajsiglova L, Kalkusova K, Strnadova K, Lacina L, Dvorankova B, Novotny J, Kolar M, et al. 2023. Novel PD-L1- and collagen-expressing patient-derived cell line of undifferentiated pleomorphic sarcoma (JBT19) as a model for cancer immunotherapy. Scientific Reports. 13(1).
Application
- Each vial contains > 1X106 viable cells.
- Cells are tested negative for infectious diseases by an Essential CLEAR Panel by Charles River Animal Diagnostic Services.
- Cells are verified to be of human origin and negative for interspecies contamination from mouse, rat, Chinese hamster, Golden Syrian hamster, and nonhuman primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
- Cells are negative for mycoplasma contamination.
Features and Benefits
JBT-19 undifferentiated pleomorphic sarcoma cells simultaneously express PD-L1 and collagen and are used for immunotherapeutic and cancer research.
Preparation Note
JBT-19 cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages without significantly affecting cell marker expression and function.
Other Notes
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@milliporesigma.com.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Lagerklasse
10 - Combustible liquids
WGK
WGK 2
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.