Merck
BR701430
BRAND® Life Science heat sealing film
suitable for PCR, storage, aluminum film (adhesive backed), operating temperature (-20)—120 °C, pack of 100 ea
Synonym(e):
aluminum film, cover, plate film, seal, sealing film
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
UNSPSC Code:
41122410
NACRES:
NB.43
eCl@ss:
32120613
Manufacturer/tradename:
BRAND 701430
Material:
aluminum film (adhesive backed)
Sterility:
non-sterile
Packaging:
pack of 100 ea
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
aluminum film (adhesive backed)
sterility
non-sterile
feature
DNA free, RNase and DNase free
packaging
pack of 100 ea
manufacturer/tradename
BRAND 701430
parameter
(-20)—120 °C operating temperature
suitability
suitable for PCR
General description
Heat sealing films provide protection against evaporation and contamination. Aluminum foils are used for PP and PS plates. They are resistant to solvents and are used both at low temperatures and for storage at room temperature. Depending on the application, they are peelable or pierceable.
Features and Benefits
  • Tight seal for minimizing evaporation
  • DMSO- and solvent-resistant
  • Peelable and pierceable
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)