Merck
BR701431
BRAND® Life Science heat sealing film
qPCR, suitable for PCR, storage, PE/PP/adhesive, operating temperature (-80)—110 °C, pack of 100 ea, transparent
Synonym(e):
cover, plate film, seal, sealing film
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
UNSPSC Code:
41122410
NACRES:
NB.43
eCl@ss:
32120613
Manufacturer/tradename:
BRAND 701431
Material:
polyethylene film, polypropylene film (adhesive backed)
Sterility:
non-sterile
Packaging:
pack of 100 ea
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polyethylene film, polypropylene film (adhesive backed)
sterility
non-sterile
feature
DNA free, RNase and DNase free, qPCR
packaging
pack of 100 ea
manufacturer/tradename
BRAND 701431
parameter
(-80)—110 °C operating temperature
suitability
suitable for PCR
General description
Heat sealing films provide protection against evaporation and contamination. Aluminum foils are used for PP and PS plates. They are resistant to solvents and are used both at low temperatures and for storage at room temperature. Depending on the application, they are peelable or pierceable.
Features and Benefits
  • Tight seal for minimizing evaporation
  • DMSO- and solvent-resistant
  • Peelable and pierceable
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)