Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
NACRES:
NA.81
UNSPSC Code:
41106514
eCl@ss:
32150313
Biological source:
human bronchi
Morphology:
(smooth muscle)
Relevant disease(s):
asthma
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenbiological source
human bronchi
packaging
pkg of 500,000 cells
morphology
(smooth muscle)
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
asthma
shipped in
ambient
storage temp.
room temp
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
Application
Primary Human Bronchial Smooth Muscle Cells (HBSMC) are isolated from the bronchi from single donors and are provided in a proliferating format. HBSMC stain positive for smooth muscle α-actin. They connect the endings of the bronchial cartilage and consequently regulate the airway lumen. Under pathological conditions like asthma, the amount of smooth muscle cells in the bronchi walls is increased leading to smooth muscle hypertrophy or hyperplasia. Hence, HBSMC are suitable for in vitro studies of asthma and other pulmonary diseases.
Biochem/physiol Actions
Bronchi
Preparation Note
Click here for more information.
Analysis Note
Rigid quality control tests are performed for each lot of Human Smooth Muscle Cells. They are tested for cell morphology, adherence rate, and cell viability. Flow cytometric analyses for cell-type specific markers, e.g. smooth muscle a-actin, are carried out for each lot. Growth performance is tested through multiple passages up to 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).
Other Notes
Recommended Plating Density: 7500 - 10000 cells per cm2Passage After Thawing: P2Tested Markers: Smooth muscle alpha-actin positiveGuaranteed population doublings: >15
Recommended Primary Cell Culture Media:Link
Disclaimer
Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
This product is regulated in France when intended for scientific purposes, including import, export, and conservation activities (Articles L.1211-1, paragraph 2, and L.1243-4 of the French Public Health Code). The purchaser (i.e. end user) is required to obtain an import authorization from the France Ministry responsible for Research (as referenced in Article L.1245-5-1 II of the French Public Health Code) or a conservation authorization from the same ministry (as referenced in Article L.1243-4 of the French Public Health Code). By placing an order for this product, you confirm that you have obtained the appropriate import or conservation authorization for the product. For more information regarding the use of human biological samples for research purposes in France, please contact the relevant French authorities.
Still not finding the right product?
Explore all of our products under Human Bronchial Smooth Muscle Cells (HBSMC)
Lagerklasse
12 - Non Combustible Liquids
WGK
WGK 3
Flammpunkt (°F)
No data available
Flammpunkt (°C)
No data available
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.