Fortfahren mit
MTCM4017
MTC™ Bio Inoculating Loops and Needles
rigid loop, capacity 1 μL, polystyrene, sterile, case of 1000 ea (50 peel packs of 20ea)
Synonym(e):
culture needle, cutlure loop, MTC Bio M4017, sampling loop, sampling tool
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41161502
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polystyrene
sterility
sterile
packaging
case of 1000 ea (50 peel packs of 20ea)
manufacturer/tradename
MTC™ Bio M4017
capacity
1 μL
General description
Sterile disposable plastic loops and needles designed for use in quantitative procedures such as sampling, urine counts, and serial dilutions, as well as for bacterial inoculation. Because they are pre-sterilized and designed for single use, there is no risk of cross-contamination and no need for flaming. The unique hexagonal shape of the shaft makes it easier to grip, orientate, and guide through a smooth streaking process. The loop surfaces are extremely smooth with no rough edges and no flashing or burrs on the loop head. This results in the elimination of cutting or gouging of the agar surface.
Flexible Polypropylene Loops - Make it easier to maneuver them within Petri dishes, test tubes, or other confined spaces, improving the researcher′s ability to collect and transfer samples efficiently. The flexibility allows for more precise sampling from cultures, especially when working with small or delicate colonies that could be easily damaged by more rigid instruments.
Rigid Polystyrene Loops & Needles - Allow for more precise streaking patterns on agar plates, which is essential for isolating individual colonies or performing quantitative assays. They are also better suited for handling viscous or dense samples, where flexibility might impede the ability to effectively collect and transfer the sample.
Features and Benefits
  • Sterilized to SAL 10-6
  • Easy-grip hexagonal handle design
  • Flexible PP or Rigid PS available
  • Individually wrapped or peel packs of 20
Legal Information
MTC is a trademark of MTC Bio
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
MTCM4017WMTCM4035MTCM4035W
description
rigid loop, capacity 1 μL, polystyrene, sterile, case of 1000 ea (50 peel packs of 20ea)
description
rigid loop, capacity 1 μL, polystyrene, sterile, case of 1000 ea (individually wrapped)
description
flexible loop, capacity 5 μL, polypropylene, sterile, case of 1000 ea (50 peel packs of 20ea)
description
flexible loop, capacity 5 μL, polypropylene, sterile, case of 1000 ea (individually wrapped)
material
polystyrene
material
polystyrene
material
polypropylene
material
polypropylene
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
capacity
1 μL
capacity
1 μL
capacity
5 μL
capacity
5 μL
manufacturer/tradename
MTC™ Bio M4017
manufacturer/tradename
MTC™ Bio M4017-W
manufacturer/tradename
MTC™ Bio M4035
manufacturer/tradename
MTC™ Bio M4035-W
packaging
case of 1000 ea (50 peel packs of 20ea)
packaging
case of 1000 ea (individually wrapped)
packaging
case of 1000 ea (50 peel packs of 20ea)
packaging
case of 1000 ea (individually wrapped)
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)