Direkt zum Inhalt
Merck

ZRB3045

Anti-HNRNPA2B1 Antibody, clone 1J11 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells

Nachhaltigkeitsmerkmale

enhanced validation

Verbesserte Validierung

Synonym(e):

HNRPA2B1, Heterogeneous nuclear ribonucleoproteins A2/B1

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern

Über diesen Artikel

UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
1J11, recombinant monoclonal
Application:
ICC, IHC, WB, affinity binding assay
Citations:
-
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

rabbit

Quality Segment

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J11, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 37.43 kDa, observed mol wt ~37.00 kDa

purified by

using Protein A

species reactivity

human, rat

species reactivity (predicted by homology)

mouse, monkey, bovine

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Sustainability features

concentration

0.3 mg/mL

technique(s)

affinity binding assay: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

General description

Heterogeneous nuclear ribonucleoproteins A2/B1 (UniProt: P22626; also known as hnRNP A2/B1) is encoded by the HNRNPA2B1 (also known as HNRPA2B1) gene (Gene ID: 3181) in human. HNRNPA2B1 plays a critical role in various aspects of RNA metabolism, including splicing, transport, stability, and translation. HNRNPA2B1 contains two RNA recognition motifs (RRMs) that facilitate binding to RNA sequences, enabling it to influence the processing and regulation of mRNA precursors and other RNA molecules. It is ubiquitously expressed in most tissues, with higher levels observed in the brain, where it is involved in neuronal RNA processing and regulation. HNRNPA2B1 is predominantly localized in the nucleus, where it associates with heterogeneous nuclear RNA (hnRNA) to form ribonucleoprotein complexes, although it can shuttle between the nucleus and the cytoplasm to execute its functions. Dysregulation of HNRNPA2B1 is implicated in several diseases, particularly neurodegenerative disorders such as amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD). HNRNPA2B1 has also been linked to cancer; altered expression affecting tumor growth, metastasis, and response to therapy. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.
We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal half of human HNRNPA2B1.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK-293 cell lysate.

Western Blotting Analysis: A 1:20,000 dilution of this antibody detected HNRNPA2B1 in HEK-293 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:20,000 dilution from a representative lot detected HNRNPA2B1 in K-562 cell lysate and a 1:10,000 dilution in NIH/3T3 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HNRNPA2B1 in HEK-293 cells.

Affinity Binding Assay: A representative lot of this antibody bound with HNRNPA2B1 peptide with at least one hundred thousand-fold (100,000X) higher affinity than with non-specific control peptide.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected HNRNPA2B1 in human small intestine and rat testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 1J11 is a ZooMAb® rabbit recombinant monoclonal antibody that detects HNRNPA2B1. It targets an epitope within 19 amino acids from the N-terminal half.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Probieren Sie unser Produkt-Auswahlhilfe, um Ihre Auswahl einzugrenzen


Lagerklasse

11 - Combustible Solids

Was ist los?

WGK 1

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

Die passende Version wird nicht angezeigt?

Wenn Sie eine bestimmte Version benötigen, können Sie anhand der Lot- oder Chargennummer nach einem spezifischen Zertifikat suchen.

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen