Skip to Content
Merck

AABH93DE453E

AA BLOCKS, INC.

Hydroxypyruvic acid

Synonym(s):

(2E)-2,3-dihydroxy-2-propenoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C3H4O4
CAS Number:
Molecular Weight:
104.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C3H4O4/c4-1-2(5)3(6)7/h4H,1H2,(H,6,7)

Inchi Key

HHDDCCUIIUWNGJ-UHFFFAOYSA-N

Purity

95%

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library