Skip to Content
Merck

AABH97CD7BB4

AA BLOCKS, INC.

Flufenpyr-ethyl

Synonym(s):

ethyl [2-chloro-4-fluoro-5-(5-methyl-6-oxo-4-(trifluoromethyl)-1(6H)-pyridazinyl)phenoxy]acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H13O4N2Cl1F4
CAS Number:
Molecular Weight:
408.74
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Storage temp.

N/A

Inchi Code

1S/C16H13ClF4N2O4/c1-3-26-14(24)7-27-13-5-12(11(18)4-10(13)17)23-15(25)8(2)9(6-22-23)16(19,20)21/h4-6H,3,7H2,1-2H3

Inchi Key

DNUAYCRATWAJQE-UHFFFAOYSA-N

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library