Skip to Content
Merck

AABH97CEC8BE

AA BLOCKS, INC.

4-(4-Bromophenyl)-N-(3-chlorophenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydropyrimidine-5-carboxamide

Synonym(s):

4-(4-bromophenyl)-N-(3-chlorophenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C18H15O1N3Cl1Br1S1
CAS Number:
Molecular Weight:
436.76
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C18H15BrClN3OS/c1-10-15(17(24)22-14-4-2-3-13(20)9-14)16(23-18(25)21-10)11-5-7-12(19)8-6-11/h2-9,16H,1H3,(H,22,24)(H2,21,23,25)

Inchi Key

UKZLBNIWRMNBOK-UHFFFAOYSA-N

Purity

90%

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library