Skip to Content
Merck

AABH97D2D4E0

AA BLOCKS, INC.

Betamethasone 21-Valerate

Synonym(s):

(11beta,16beta)-9-fluoro-11,17-dihydroxy-16-methyl-3,20-dioxopregna-1,4-dien-21-yl pentanoate

Select a Size

Change View

About This Item

Linear Formula:
C27H37O6F1
CAS Number:
Molecular Weight:
476.59
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C27H37FO6/c1-5-6-7-23(32)34-15-22(31)27(33)16(2)12-20-19-9-8-17-13-18(29)10-11-24(17,3)26(19,28)21(30)14-25(20,27)4/h10-11,13,16,19-21,30,33H,5-9,12,14-15H2,1-4H3/t16-,19-,20-,21-,24-,25-,26-,27-/m0/s1

Inchi Key

FEROCCAEIIKMJT-SUYDQAKGSA-N

Purity

98%

Storage temp.

N/A

Country of Origin

US



Still not finding the right product?

Explore all of our products under Betamethasone 21-Valerate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library