Skip to Content
Merck

AABH97D2E2D3

AA BLOCKS, INC.

Dynasore hydrate

Synonym(s):

N'-[(E)-(3,4-dihydroxyphenyl)methylidene]-3-hydroxy-2-naphthohydrazide hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C18H16O5N2
CAS Number:
Molecular Weight:
340.34
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C18H14N2O4.H2O/c21-15-6-5-11(7-17(15)23)10-19-20-18(24)14-8-12-3-1-2-4-13(12)9-16(14)22;/h1-10,21-23H,(H,20,24);1H2/b19-10+;

Inchi Key

FKWZHQSDBMSHTN-ZIOFAICLSA-N

Storage temp.

N/A

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library