Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
25 mg | Ships in 21 days - from Aldrich Partner.
| € 165,22 |
50 mg | Ships in 21 days - from Aldrich Partner.
| € 237,22 |
5 mg | Ships in 21 days - from Aldrich Partner.
| € 921,16 |
About This Item
Vendor SKU: AA01ENUX-25MG
€ 165,22
Ships in 21 days - from Aldrich Partner.
Inchi Code
1S/C35H57N15O9/c1-19(45-31(55)25(13-21-15-41-18-44-21)48-29(53)22(37)12-20-14-40-17-43-20)28(52)49-26(16-51)33(57)50-11-5-8-27(50)32(56)46-23(7-4-10-42-35(38)39)30(54)47-24(34(58)59)6-2-3-9-36/h14-15,17-19,22-27,51H,2-13,16,36-37H2,1H3,(H,40,43)(H,41,44)(H,45,55)(H,46,56)(H,47,54)(H,48,53)(H,49,52)(H,58,59)(H4,38,39,42)/t19-,22-,23-,24-,25-,26-,27-/m0/s1
Inchi Key
MSJILSSAIKNNOL-YJJSATQUSA-N
Purity
99%
Country of Origin
CN
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.