Skip to Content
Merck

271543

cis-8,11,14-Eicosatrienoic acid

99%

Synonym(s):

Homo-γ-linolenic acid

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Linear Formula:
CH3(CH2)3(CH2CH=CH)3(CH2)6CO2H
CAS Number:
Molecular Weight:
306.48
Beilstein:
1796533
EC Number:
MDL number:
UNSPSC Code:
12352103
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

99%

refractive index

n20/D 1.478 (lit.)

SMILES string

CCCCC\C=C/C\C=C/C\C=C/CCCCCCC(O)=O

InChI

1S/C20H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13H,2-5,8,11,14-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-

InChI key

HOBAELRKJCKHQD-QNEBEIHSSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service