Skip to Content
Merck

Skip To

856444

(+)-Griseofulvin

Synonym(s):

(2S)-trans-7-Chloro-2′,4,6-trimethoxy-6′-methylspiro(benzofuran-2[3H],1′-[2]cyclohexene)-3,4′-dione, Griseofulvin

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C17H17ClO6
CAS Number:
Molecular Weight:
352.77
UNSPSC Code:
12352103
EC Number:
204-767-4
Beilstein/REAXYS Number:
95226
MDL number:


optical activity

[α]17/D +370°, c = 1 in chloroform

mp

218-220 °C (lit.)

SMILES string

COc1cc(OC)c2C(=O)[C@]3(Oc2c1Cl)[C@H](C)CC(=O)C=C3OC

Features and Benefits

Antimitotic.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library