Skip to Content
Merck

Skip To

86355

TentaGel MB-COOH

extent of labeling: ~0.40 mmol/g loading

Synonym(s):

O-(2-Succinylaminoethyl)polyethylene glycol polymer-bound

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:


extent of labeling

~0.40 mmol/g loading

particle size

140-170 μm

storage temp.

2-8°C

Legal Information

TentaGel is a trademark of Rapp Polymere GmbH




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library