Skip to Content
Merck

Skip To

GF42657150

Nickel-chromium alloy, Ni80Cr20

100m wire, 0.30mm diameter, annealed

Synonym(s):

NI055135

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 382,00

About This Item

NACRES:
NA.23
UNSPSC Code:
11101711

€ 382,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

wire

composition

Ni80Cr20

reaction suitability

core: chromium, core: nickel

manufacturer/tradename

Goodfellow 426-571-50

L × diam.

100 m × 0.30 mm

application(s)

metallization

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF44613323GF10672253GF34032259
description

100m wire, 0.30mm diameter, annealed

description

25m wire, 1.0mm diameter, hard

description

1000m wire, 0.05mm diameter, hard

description

2000m wire, 0.05mm diameter, hard

form

wire

form

wire

form

wire

form

wire

manufacturer/tradename

Goodfellow 426-571-50

manufacturer/tradename

Goodfellow 446-133-23

manufacturer/tradename

Goodfellow 106-722-53

manufacturer/tradename

Goodfellow 340-322-59

application(s)

metallization

application(s)

metallization

application(s)

metallization

application(s)

metallization

reaction suitability

core: nickel, core: chromium

reaction suitability

core: chromium, core: nickel

reaction suitability

core: nickel, core: chromium

reaction suitability

core: nickel, core: chromium

L × diam.

100 m × 0.30 mm

L × diam.

25 mm × 1.0 mm

L × diam.

1000 m × 0.05 mm

L × diam.

2000 m × 0.05 mm

composition

Ni80Cr20

composition

Ni80Cr20

composition

Ni80Cr20

composition

Ni80Cr20


Still not finding the right product?

Explore all of our products under Nickel-chromium alloy, Ni80Cr20




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters